Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATTTTATAGGCATAGACGATAGG[A/G]TTCATCATGGAGTTGGCATGGGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600445 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADORA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADORA3 - adenosine A3 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000677.3 | 1601 | Silent Mutation | AAC,AAT | N,N 278 | NP_000668.1 | |
NM_001302678.1 | 1601 | UTR 3 | NP_001289607.1 | |||
NM_001302679.1 | 1601 | Silent Mutation | AAC,AAT | N,N 133 | NP_001289608.1 |
TMIGD3 - transmembrane and immunoglobulin domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081976.2 | 1601 | Intron | NP_001075445.1 | |||
NM_001302680.1 | 1601 | Intron | NP_001289609.1 | |||
NM_020683.6 | 1601 | Intron | NP_065734.5 |