Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCCAAGGTGGGTATCCTCACACT[C/T]CTTCAGCTTGATGAGAGCTGTGACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602651 MIM: 606737 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NRDC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NRDC - nardilysin convertase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101662.1 | 3398 | Missense Mutation | AAG,GAG | K,E 1026 | NP_001095132.1 | |
NM_001242361.1 | 3398 | Missense Mutation | AAG,GAG | K,E 962 | NP_001229290.1 | |
NM_002525.2 | 3398 | Missense Mutation | AAG,GAG | K,E 1094 | NP_002516.2 | |
XM_005270903.1 | 3398 | Missense Mutation | AAG,GAG | K,E 789 | XP_005270960.1 | |
XM_011541522.1 | 3398 | Missense Mutation | AAG,GAG | K,E 984 | XP_011539824.1 | |
XM_011541523.1 | 3398 | Missense Mutation | AAG,GAG | K,E 894 | XP_011539825.1 | |
XM_011541525.2 | 3398 | Missense Mutation | AAG,GAG | K,E 789 | XP_011539827.1 | |
XM_017001375.1 | 3398 | Missense Mutation | AAG,GAG | K,E 894 | XP_016856864.1 |
OSBPL9 - oxysterol binding protein like 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |