Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCTAATCTTCCTATTGGGATTCC[A/G]TATTCAATTTCCTTTAAGAGGTATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 615843 | ||||||||||||||||||||
Literature Links: |
DEGS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DEGS1 - delta 4-desaturase, sphingolipid 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321541.1 | 486 | Silent Mutation | CCA,CCG | P,P 119 | NP_001308470.1 | |
NM_001321542.1 | 486 | Silent Mutation | CCA,CCG | P,P 83 | NP_001308471.1 | |
NM_003676.3 | 486 | Silent Mutation | CCA,CCG | P,P 119 | NP_003667.1 | |
XM_017002648.1 | 486 | Silent Mutation | CCA,CCG | P,P 83 | XP_016858137.1 |