Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGTTTTTCCACACCCAATTACAG[G/T]AGAAGAGCACCCCTATATCCAGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606909 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
VAMP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VAMP4 - vesicle associated membrane protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185127.1 | 4709 | UTR 3 | NP_001172056.1 | |||
NM_003762.4 | 4709 | UTR 3 | NP_003753.2 | |||
XM_017002713.1 | 4709 | UTR 3 | XP_016858202.1 |