Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGAAGTTAAATGAAATACCTCCGG[G/T]GAGGGCAGGCCGCAGGGAGGCTCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C1orf106 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C1orf106 - chromosome 1 open reading frame 106 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142569.2 | 80 | Intron | NP_001136041.1 | |||
NM_018265.3 | 80 | Missense Mutation | GGG,GTG | G,V 27 | NP_060735.3 | |
XM_011509754.2 | 80 | Intron | XP_011508056.1 | |||
XM_011509755.1 | 80 | Intron | XP_011508057.1 | |||
XM_011509756.2 | 80 | Missense Mutation | GGG,GTG | G,V 27 | XP_011508058.1 |