Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACAAACTGAAATGGTGCTGTAAAT[C/T]TGAAGACAATCTAAATAGAAAAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 610471 | ||||||||||||||||||||
Literature Links: |
ANGEL2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANGEL2 - angel homolog 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300753.1 | 1483 | Missense Mutation | AAT,GAT | N,D 318 | NP_001287682.1 | |
NM_001300755.1 | 1483 | Missense Mutation | AAT,GAT | N,D 318 | NP_001287684.1 | |
NM_001300757.1 | 1483 | Missense Mutation | AAT,GAT | N,D 275 | NP_001287686.1 | |
NM_001300758.1 | 1483 | Missense Mutation | AAT,GAT | N,D 275 | NP_001287687.1 | |
NM_144567.4 | 1483 | Missense Mutation | AAT,GAT | N,D 444 | NP_653168.2 | |
XM_005273344.1 | 1483 | Missense Mutation | AAT,GAT | N,D 422 | XP_005273401.1 | |
XM_005273345.1 | 1483 | Missense Mutation | AAT,GAT | N,D 422 | XP_005273402.1 | |
XM_005273346.2 | 1483 | Missense Mutation | AAT,GAT | N,D 422 | XP_005273403.1 | |
XM_005273347.3 | 1483 | Missense Mutation | AAT,GAT | N,D 365 | XP_005273404.1 | |
XM_017002774.1 | 1483 | Missense Mutation | AAT,GAT | N,D 422 | XP_016858263.1 | |
XM_017002775.1 | 1483 | Missense Mutation | AAT,GAT | N,D 318 | XP_016858264.1 | |
XM_017002776.1 | 1483 | Missense Mutation | AAT,GAT | N,D 318 | XP_016858265.1 | |
XM_017002777.1 | 1483 | Missense Mutation | AAT,GAT | N,D 318 | XP_016858266.1 | |
XM_017002778.1 | 1483 | Missense Mutation | AAT,GAT | N,D 318 | XP_016858267.1 |
VASH2 - vasohibin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |