Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTGGTTACAGGTACCATTGTTG[G/T]ATACTGGTGACATCATCATTGACGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 172200 | ||||||||||||||||||||
Literature Links: |
PGD PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PGD - phosphogluconate dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304451.1 | 403 | Intron | NP_001291380.1 | |||
NM_001304452.1 | 403 | Missense Mutation | GAT,TAT | D,Y 80 | NP_001291381.1 | |
NM_002631.3 | 403 | Missense Mutation | GAT,TAT | D,Y 93 | NP_002622.2 |