Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGATGCTCTCGCAGTTCTTCTCA[C/T]GGTGGCACCTACGGCCAGCGGTGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602863 | ||||||||||||||||||||
Literature Links: |
WNT9A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
WNT9A - Wnt family member 9A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003395.2 | 1515 | Missense Mutation | CAT,CGT | H,R 315 | NP_003386.1 | |
XM_011544271.2 | 1515 | Missense Mutation | CAT,CGT | H,R 245 | XP_011542573.1 |