Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTACTCTACAGTATATGACATTTA[A/G]TATGATCAACTTTTGATATACAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602549 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PKN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PKN2 - protein kinase N2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320707.1 | Intron | NP_001307636.1 | ||||
NM_001320708.1 | Intron | NP_001307637.1 | ||||
NM_001320709.1 | Intron | NP_001307638.1 | ||||
NM_006256.3 | Intron | NP_006247.1 | ||||
XM_011541772.2 | Intron | XP_011540074.1 | ||||
XM_017001782.1 | Intron | XP_016857271.1 | ||||
XM_017001783.1 | Intron | XP_016857272.1 |
PKN2-AS1 - PKN2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |