Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGATTAACATCCTGTCGGGAGA[A/G]ACTGTCAAAGCTGGGGACAGGGACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611632 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBIAD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UBIAD1 - UbiA prenyltransferase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013319.2 | 385 | Silent Mutation | GAA,GAG | E,E 17 | NP_037451.1 | |
XM_006710590.3 | 385 | Silent Mutation | GAA,GAG | E,E 17 | XP_006710653.1 | |
XM_011541304.2 | 385 | Silent Mutation | GAA,GAG | E,E 17 | XP_011539606.1 |