Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACCCAGGGGGCCCTTGGCGGAGG[C/T]GGGCAATTTAACAACTGGTCTGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610036 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLDN19 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLDN19 - claudin 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001123395.1 | 837 | UTR 3 | NP_001116867.1 | |||
NM_001185117.1 | 837 | UTR 3 | NP_001172046.1 | |||
NM_148960.2 | 837 | Missense Mutation | ACC,GCC | T,A 216 | NP_683763.2 |