Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTCTTCTTATTTTTTCATCTCGA[A/G]TGGTTTCTTCTAACTCCGGTGGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615354 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRIF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRIF1 - ligand dependent nuclear receptor interacting factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006945.1 | 2731 | Missense Mutation | ACT,ATT | T,I 204 | NP_001006946.1 | |
NM_018372.3 | 2731 | Missense Mutation | ACT,ATT | T,I 740 | NP_060842.3 | |
XM_005271029.3 | 2731 | Missense Mutation | ACT,ATT | T,I 739 | XP_005271086.1 | |
XM_011541768.1 | 2731 | Missense Mutation | ACT,ATT | T,I 713 | XP_011540070.1 | |
XM_017001768.1 | 2731 | Missense Mutation | ACT,ATT | T,I 712 | XP_016857257.1 | |
XM_017001769.1 | 2731 | Intron | XP_016857258.1 |