Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCATCTCTTCATTAGCACATAAGC[A/G]GTTCAACTTGGGATACGCAAGGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612178 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM102B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM102B - family with sequence similarity 102 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HENMT1 - HEN1 methyltransferase homolog 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102592.1 | 1481 | Missense Mutation | CGC,TGC | R,C 354 | NP_001096062.1 | |
NM_144584.2 | 1481 | Missense Mutation | CGC,TGC | R,C 354 | NP_653185.2 | |
XM_005270411.1 | 1481 | Missense Mutation | CGC,TGC | R,C 362 | XP_005270468.1 | |
XM_017000179.1 | 1481 | Missense Mutation | CGC,TGC | R,C 198 | XP_016855668.1 |