Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGCAGAACTCACAGATGTAGTCCC[A/G]GGTGTCTGCAGGCATATGAGGGACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF672 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF672 - zinc finger protein 672 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF692 - zinc finger protein 692 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136036.2 | 1370 | Missense Mutation | CGG,TGG | R,W 392 | NP_001129508.1 | |
NM_001193328.1 | 1370 | Missense Mutation | CGG,TGG | R,W 342 | NP_001180257.1 | |
NM_017865.3 | 1370 | Missense Mutation | CGG,TGG | R,W 387 | NP_060335.2 | |
XM_011544220.1 | 1370 | Missense Mutation | CGG,TGG | R,W 538 | XP_011542522.1 | |
XM_011544221.1 | 1370 | Missense Mutation | CGG,TGG | R,W 537 | XP_011542523.1 | |
XM_011544222.1 | 1370 | Missense Mutation | CGG,TGG | R,W 387 | XP_011542524.1 | |
XM_011544223.2 | 1370 | Missense Mutation | CGG,TGG | R,W 493 | XP_011542525.1 | |
XM_011544227.1 | 1370 | Missense Mutation | CGG,TGG | R,W 360 | XP_011542529.1 | |
XM_017001698.1 | 1370 | Missense Mutation | CGG,TGG | R,W 386 | XP_016857187.1 | |
XM_017001699.1 | 1370 | Missense Mutation | CGG,TGG | R,W 512 | XP_016857188.1 | |
XM_017001700.1 | 1370 | Missense Mutation | CGG,TGG | R,W 361 | XP_016857189.1 | |
XM_017001701.1 | 1370 | Missense Mutation | CGG,TGG | R,W 386 | XP_016857190.1 | |
XM_017001702.1 | 1370 | Missense Mutation | CGG,TGG | R,W 361 | XP_016857191.1 | |
XM_017001703.1 | 1370 | Missense Mutation | CGG,TGG | R,W 183 | XP_016857192.1 | |
XM_017001704.1 | 1370 | Missense Mutation | CGG,TGG | R,W 183 | XP_016857193.1 | |
XM_017001705.1 | 1370 | Missense Mutation | CGG,TGG | R,W 183 | XP_016857194.1 |