Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCTCCTTCACGTCATCTTTGTGT[C/G]ACAGCAGGACATCCGGGTTTGGAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FAM129A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM129A - family with sequence similarity 129 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052966.3 | 3711 | Nonsense Mutation | TCA,TGA | S,* 906 | NP_443198.1 | |
XM_011509140.2 | 3711 | Nonsense Mutation | TCA,TGA | S,* 965 | XP_011507442.1 | |
XM_011509141.2 | 3711 | Nonsense Mutation | TCA,TGA | S,* 928 | XP_011507443.1 |