Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCCCAATTCTCTTTAGCTGCCTC[C/T]TCACAGCCTCCTTGATCTCCTTGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
OR6K6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OR6K6 - olfactory receptor family 6 subfamily K member 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OR6N1 - olfactory receptor family 6 subfamily N member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005185.1 | 1137 | Missense Mutation | AAG,AGG | K,R 302 | NP_001005185.1 | |
XM_017000325.1 | 1137 | Missense Mutation | AAG,AGG | K,R 302 | XP_016855814.1 | |
XM_017000326.1 | 1137 | Missense Mutation | AAG,AGG | K,R 302 | XP_016855815.1 | |
XM_017000327.1 | 1137 | Missense Mutation | AAG,AGG | K,R 302 | XP_016855816.1 |