Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGTAGAGCACGCGCCCATTGATG[C/T]GGTCACGGATCTGCTCCACACGCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 614994 MIM: 150310 | ||||||||||||||||||||
Literature Links: |
CAMK1G PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CAMK1G - calcium/calmodulin dependent protein kinase IG | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LAMB3 - laminin subunit beta 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000228.2 | 3623 | Missense Mutation | CAC,CGC | H,R 1160 | NP_000219.2 | |
NM_001017402.1 | 3623 | Missense Mutation | CAC,CGC | H,R 1160 | NP_001017402.1 | |
NM_001127641.1 | 3623 | Missense Mutation | CAC,CGC | H,R 1160 | NP_001121113.1 | |
XM_005273124.4 | 3623 | Missense Mutation | CAC,CGC | H,R 1160 | XP_005273181.1 | |
XM_017001272.1 | 3623 | Missense Mutation | CAC,CGC | H,R 1096 | XP_016856761.1 |
MIR4260 - microRNA 4260 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |