Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGCCAGGCTCTATTTCCAGCGCC[C/T]GGTGACTTTAGCCTTCCCGCGGGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 191045 | ||||||||||||||||||||
Literature Links: |
TNNT2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TNNT2 - troponin T2, cardiac type | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000364.3 | 1070 | Missense Mutation | AGG,GGG | R,G 292 | NP_000355.2 | |
NM_001001430.2 | 1070 | Missense Mutation | AGG,GGG | R,G 285 | NP_001001430.1 | |
NM_001001431.2 | 1070 | Missense Mutation | AGG,GGG | R,G 282 | NP_001001431.1 | |
NM_001001432.2 | 1070 | Missense Mutation | AGG,GGG | R,G 279 | NP_001001432.1 | |
NM_001276345.1 | 1070 | Missense Mutation | AGG,GGG | R,G 295 | NP_001263274.1 | |
NM_001276346.1 | 1070 | Missense Mutation | AGG,GGG | R,G 252 | NP_001263275.1 | |
NM_001276347.1 | 1070 | Missense Mutation | AGG,GGG | R,G 285 | NP_001263276.1 | |
XM_006711508.3 | 1070 | Missense Mutation | AGG,GGG | R,G 285 | XP_006711571.1 | |
XM_006711509.3 | 1070 | Missense Mutation | AGG,GGG | R,G 284 | XP_006711572.1 | |
XM_011509938.2 | 1070 | Missense Mutation | AGG,GGG | R,G 295 | XP_011508240.1 | |
XM_011509939.1 | 1070 | Missense Mutation | AGG,GGG | R,G 294 | XP_011508241.1 | |
XM_011509940.2 | 1070 | Missense Mutation | AGG,GGG | R,G 294 | XP_011508242.1 | |
XM_011509941.2 | 1070 | Missense Mutation | AGG,GGG | R,G 293 | XP_011508243.1 | |
XM_011509942.2 | 1070 | Missense Mutation | AGG,GGG | R,G 280 | XP_011508244.1 | |
XM_011509943.2 | 1070 | Missense Mutation | AGG,GGG | R,G 280 | XP_011508245.1 | |
XM_011509944.2 | 1070 | Missense Mutation | AGG,GGG | R,G 279 | XP_011508246.1 | |
XM_011509946.1 | 1070 | Missense Mutation | AGG,GGG | R,G 226 | XP_011508248.1 | |
XM_017002216.1 | 1070 | Missense Mutation | AGG,GGG | R,G 284 | XP_016857705.1 | |
XM_017002217.1 | 1070 | Missense Mutation | AGG,GGG | R,G 282 | XP_016857706.1 |