Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCCCGTCCGTCATCCCGGTCTCC[C/T]GTCTCATAGGATCCCCCGGCTGGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 600663 | ||||||||||||||||||||
Literature Links: |
MEF2D PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MEF2D - myocyte enhancer factor 2D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271629.1 | 1907 | Silent Mutation | ACA,ACG | T,T 473 | NP_001258558.1 | |
NM_005920.3 | 1907 | Silent Mutation | ACA,ACG | T,T 480 | NP_005911.1 | |
XM_005245169.4 | 1907 | Silent Mutation | ACA,ACG | T,T 480 | XP_005245226.1 | |
XM_005245170.3 | 1907 | Silent Mutation | ACA,ACG | T,T 480 | XP_005245227.1 | |
XM_006711330.3 | 1907 | Silent Mutation | ACA,ACG | T,T 480 | XP_006711393.1 | |
XM_006711332.3 | 1907 | Silent Mutation | ACA,ACG | T,T 479 | XP_006711395.1 | |
XM_006711333.2 | 1907 | Silent Mutation | ACA,ACG | T,T 473 | XP_006711396.1 | |
XM_006711334.3 | 1907 | Silent Mutation | ACA,ACG | T,T 472 | XP_006711397.1 | |
XM_011509569.2 | 1907 | Silent Mutation | ACA,ACG | T,T 480 | XP_011507871.1 | |
XM_017001314.1 | 1907 | Silent Mutation | ACA,ACG | T,T 473 | XP_016856803.1 | |
XM_017001315.1 | 1907 | Silent Mutation | ACA,ACG | T,T 473 | XP_016856804.1 |