Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCTAACCATCAACACCACCATCT[C/G]TGCTGGATATTGTATGACACGGGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 188540 | ||||||||||||||||||||
Literature Links: |
TSHB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TSHB - thyroid stimulating hormone beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000549.4 | 178 | Missense Mutation | TCT,TGT | S,C 47 | NP_000540.2 | |
NM_001277991.1 | 178 | Intron | NP_001264920.1 | |||
XM_011542065.2 | 178 | Missense Mutation | TCT,TGT | S,C 47 | XP_011540367.1 |