Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCACTGTAGTGTCCTTCTACTATG[A/C]ACCCTTTGCTTATACCTATGTACGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
OR2L13 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OR2L13 - olfactory receptor family 2 subfamily L member 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304535.1 | 981 | Intron | NP_001291464.1 | |||
NM_175911.3 | 981 | Intron | NP_787107.1 | |||
XM_011544169.2 | 981 | Intron | XP_011542471.1 |
OR2L2 - olfactory receptor family 2 subfamily L member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004686.2 | 981 | Missense Mutation | GAA,GCA | E,A 252 | NP_001004686.1 | |
XM_011544158.2 | 981 | Missense Mutation | GAA,GCA | E,A 252 | XP_011542460.1 |