Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATACGAAATGGATTTTCTGAGATA[C/G]GGGGCAGTGTGGTACGTAAGCGGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605619 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL20 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL20 - interleukin 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018724.3 | 618 | Missense Mutation | CGG,GGG | R,G 50 | NP_061194.2 | |
XM_005273152.3 | 618 | Missense Mutation | CGG,GGG | R,G 135 | XP_005273209.1 |