Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCTGAGTGGCCCACTCAGGGTCC[A/G]GGACAGGCATACTCATCGGACCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LDLRAD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LDLRAD1 - low density lipoprotein receptor class A domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010978.3 | 554 | Silent Mutation | CCC,CCT | P,P 203 | NP_001010978.2 | |
NM_001276392.1 | 554 | Silent Mutation | CCC,CCT | P,P 164 | NP_001263321.1 | |
NM_001276393.1 | 554 | Silent Mutation | CCC,CCT | P,P 160 | NP_001263322.1 | |
NM_001276394.1 | 554 | Silent Mutation | CCC,CCT | P,P 114 | NP_001263323.1 | |
NM_001276395.1 | 554 | UTR 3 | NP_001263324.1 | |||
XM_011541446.1 | 554 | Silent Mutation | CCC,CCT | P,P 202 | XP_011539748.1 | |
XM_017001264.1 | 554 | Intron | XP_016856753.1 |