Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCCAGAGGGCGCTTGGAGGGCAG[C/T]GGAAGGATTCGGGCCTGGATAGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611616 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NADK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NADK - NAD kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198993.1 | 1299 | UTR 3 | NP_001185922.1 | |||
NM_001198994.1 | 1299 | UTR 3 | NP_001185923.1 | |||
NM_001198995.1 | 1299 | UTR 3 | NP_001185924.1 | |||
NM_023018.4 | 1299 | UTR 3 | NP_075394.3 | |||
XM_005244778.2 | 1299 | UTR 3 | XP_005244835.1 | |||
XM_006710837.2 | 1299 | UTR 3 | XP_006710900.1 | |||
XM_006710838.2 | 1299 | UTR 3 | XP_006710901.1 | |||
XM_006710839.1 | 1299 | UTR 3 | XP_006710902.1 | |||
XM_011542006.1 | 1299 | UTR 3 | XP_011540308.1 | |||
XM_011542007.1 | 1299 | UTR 3 | XP_011540309.1 | |||
XM_011542008.1 | 1299 | UTR 3 | XP_011540310.1 | |||
XM_011542009.2 | 1299 | UTR 3 | XP_011540311.1 | |||
XM_017002143.1 | 1299 | UTR 3 | XP_016857632.1 | |||
XM_017002144.1 | 1299 | UTR 3 | XP_016857633.1 | |||
XM_017002145.1 | 1299 | UTR 3 | XP_016857634.1 | |||
XM_017002146.1 | 1299 | UTR 3 | XP_016857635.1 |
SLC35E2 - solute carrier family 35 member E2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |