Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGATCCGATGCTGAGTATGCAAAT[A/G]AGCAGTGCTGATTTTGACAAGAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605769 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRIM33 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRIM33 - tripartite motif containing 33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015906.3 | 6179 | UTR 3 | NP_056990.3 | |||
NM_033020.2 | 6179 | UTR 3 | NP_148980.2 | |||
XM_005270936.3 | 6179 | UTR 3 | XP_005270993.1 | |||
XM_005270937.3 | 6179 | UTR 3 | XP_005270994.1 | |||
XM_011541568.2 | 6179 | UTR 3 | XP_011539870.1 | |||
XM_017001452.1 | 6179 | UTR 3 | XP_016856941.1 | |||
XM_017001453.1 | 6179 | UTR 3 | XP_016856942.1 | |||
XM_017001454.1 | 6179 | Intron | XP_016856943.1 |