Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCCTGACTTCTGGGACTTCTGCC[A/T]CGGCGTGCCACCCCCTTTTCCCCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 616064 | ||||||||||||||||||||
Literature Links: |
TINAGL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TINAGL1 - tubulointerstitial nephritis antigen like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204414.1 | 404 | Missense Mutation | CAC,CTC | H,L 93 | NP_001191343.1 | |
NM_001204415.1 | 404 | Intron | NP_001191344.1 | |||
NM_022164.2 | 404 | Missense Mutation | CAC,CTC | H,L 93 | NP_071447.1 | |
XM_005271106.2 | 404 | Missense Mutation | CAC,CTC | H,L 93 | XP_005271163.1 | |
XM_005271107.2 | 404 | Intron | XP_005271164.1 | |||
XM_011541946.1 | 404 | Intron | XP_011540248.1 |