Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAGCGCCCCCGGCCCGCCATGGC[C/G]GCGCGCCCCGGACCGCTCTGGCTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608271 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BMP8A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BMP8A - bone morphogenetic protein 8a | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181809.3 | 352 | Silent Mutation | GCC,GCG | A,A 2 | NP_861525.2 | |
XM_006710616.3 | 352 | Intron | XP_006710679.1 | |||
XM_011541381.2 | 352 | Intron | XP_011539683.1 | |||
XM_011541382.2 | 352 | Intron | XP_011539684.1 | |||
XM_017001198.1 | 352 | Intron | XP_016856687.1 |
LOC105378950 - mitogen-activated protein kinase 7-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011542544.2 | 352 | Intron | XP_011540846.1 |
MACF1 - microtubule-actin crosslinking factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |