Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTAGGGGAGGTCTCTGCCAGTGC[A/G]GGCTGGCCCTCAGCCCCGTCGCGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 613484 MIM: 604084 | ||||||||||||||||||||
Literature Links: |
SPEN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SPEN - spen family transcriptional repressor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZBTB17 - zinc finger and BTB domain containing 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242884.1 | 2585 | Silent Mutation | CCC,CCT | P,P 704 | NP_001229813.1 | |
NM_001287603.1 | 2585 | Silent Mutation | CCC,CCT | P,P 793 | NP_001274532.1 | |
NM_001287604.1 | 2585 | Silent Mutation | CCC,CCT | P,P 710 | NP_001274533.1 | |
NM_001324137.1 | 2585 | Silent Mutation | CCC,CCT | P,P 710 | NP_001311066.1 | |
NM_001324138.1 | 2585 | Silent Mutation | CCC,CCT | P,P 723 | NP_001311067.1 | |
NM_003443.2 | 2585 | Silent Mutation | CCC,CCT | P,P 786 | NP_003434.2 | |
XM_005245986.1 | 2585 | Silent Mutation | CCC,CCT | P,P 730 | XP_005246043.1 | |
XM_005245987.1 | 2585 | Silent Mutation | CCC,CCT | P,P 717 | XP_005246044.1 | |
XM_011542085.2 | 2585 | Silent Mutation | CCC,CCT | P,P 717 | XP_011540387.1 | |
XM_011542086.1 | 2585 | Silent Mutation | CCC,CCT | P,P 711 | XP_011540388.1 | |
XM_011542087.2 | 2585 | Silent Mutation | CCC,CCT | P,P 711 | XP_011540389.1 | |
XM_011542088.1 | 2585 | Silent Mutation | CCC,CCT | P,P 704 | XP_011540390.1 | |
XM_017002242.1 | 2585 | Silent Mutation | CCC,CCT | P,P 711 | XP_016857731.1 |