Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATTAGAATGGAAGGGGGAAAAAA[A/C]AAAACCATAACCCAACTTCATTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182310 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP1A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP1A1 - ATPase Na+/K+ transporting subunit alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000701.7 | 2983 | Intron | NP_000692.2 | |||
NM_001160233.1 | 2983 | Intron | NP_001153705.1 | |||
NM_001160234.1 | 2983 | Intron | NP_001153706.1 | |||
XM_017001360.1 | 2983 | UTR 5 | XP_016856849.1 | |||
XM_017001361.1 | 2983 | Intron | XP_016856850.1 |