Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATATTCTAATATACCTCCTCCTAC[C/T]GCTCTCCCCTCTGGGTTCCTGCTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR4781 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR4781 - microRNA 4781 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TCEANC2 - transcription elongation factor A N-terminal and central domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153035.2 | Intron | NP_694580.1 | ||||
XM_017000293.1 | Intron | XP_016855782.1 |
TMEM59 - transmembrane protein 59 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |