Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCACCATGCTCCCTTGTACGACA[C/T]GGCACAGTTAGTCACTGCAACATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601995 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TNR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TNR - tenascin R | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003285.2 | 4038 | Missense Mutation | ATG,GTG | M,V 1292 | NP_003276.3 | |
XM_011509949.2 | 4038 | Missense Mutation | ATG,GTG | M,V 1133 | XP_011508251.1 | |
XM_017002218.1 | 4038 | Missense Mutation | ATG,GTG | M,V 1292 | XP_016857707.1 | |
XM_017002219.1 | 4038 | Missense Mutation | ATG,GTG | M,V 1292 | XP_016857708.1 |