Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCACTCTTCCTAAGGAAGCAGCTG[C/T]CCCACCTTCTGAAATGATGTCTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611600 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RIMS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RIMS3 - regulating synaptic membrane exocytosis 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014747.2 | 6493 | UTR 3 | NP_055562.2 | |||
XM_011542479.2 | 6493 | UTR 3 | XP_011540781.1 |