Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAATCACCAAAATAAGTTAGTTC[C/T]AGGAAAACCAGGTGGGATTTATAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604950 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHTF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PHTF1 - putative homeodomain transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323041.1 | Intron | NP_001309970.1 | ||||
NM_001323042.1 | Intron | NP_001309971.1 | ||||
NM_001323043.1 | Intron | NP_001309972.1 | ||||
NM_001323044.1 | Intron | NP_001309973.1 | ||||
NM_001323045.1 | Intron | NP_001309974.1 | ||||
NM_001323046.1 | Intron | NP_001309975.1 | ||||
NM_001323047.1 | Intron | NP_001309976.1 | ||||
NM_001323048.1 | Intron | NP_001309977.1 | ||||
NM_001323049.1 | Intron | NP_001309978.1 | ||||
NM_001323050.1 | Intron | NP_001309979.1 | ||||
NM_001323051.1 | Intron | NP_001309980.1 | ||||
NM_001323052.1 | Intron | NP_001309981.1 | ||||
NM_001323053.1 | Intron | NP_001309982.1 | ||||
NM_006608.2 | Intron | NP_006599.2 |