Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGAAGGAAAGACAGGTCTGCTGG[A/G]GGGCCCGGGATGAGTACTGGAAGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 614772 | ||||||||||||||||||||
Literature Links: |
COA6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COA6 - cytochrome c oxidase assembly factor 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012985.2 | 214 | Missense Mutation | AGG,GGG | R,G 60 | NP_001013003.1 | |
NM_001206641.2 | 214 | Missense Mutation | AGG,GGG | R,G 90 | NP_001193570.2 | |
NM_001301733.1 | 214 | Missense Mutation | AGG,GGG | R,G 14 | NP_001288662.1 |
LOC101927765 - uncharacterized LOC101927765 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |