Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCTGGAGACGCATAAACCTGGGT[C/T]GGGAAAGTGCCCAGATCTGCCTGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 616791 | ||||||||||||||||||||
Literature Links: |
PGBD5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PGBD5 - piggyBac transposable element derived 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258311.1 | 3894 | UTR 3 | NP_001245240.1 | |||
XM_017002330.1 | 3894 | UTR 3 | XP_016857819.1 |