Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCATGGCCCAGGCAGCCACGTAG[C/T]GCTGCTTCAGGTCGTACTTGATGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
OLFML2B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OLFML2B - olfactomedin like 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297713.1 | 2819 | Missense Mutation | CAC,CGC | H,R 594 | NP_001284642.1 | |
NM_015441.2 | 2819 | Missense Mutation | CAC,CGC | H,R 593 | NP_056256.1 | |
XM_005245075.2 | 2819 | Missense Mutation | CAC,CGC | H,R 595 | XP_005245132.1 | |
XM_011509398.2 | 2819 | Missense Mutation | CAC,CGC | H,R 353 | XP_011507700.1 | |
XM_017000967.1 | 2819 | Intron | XP_016856456.1 |