Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCCTTTCCCTGAGCCTCAGGGGA[A/G]GTCCCAGGTCCTGAGCTATAGCCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 142230 | ||||||||||||||||||||
Literature Links: |
CD34 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD34 - CD34 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025109.1 | 1488 | Silent Mutation | ACC,ACT | T,T 345 | NP_001020280.1 | |
NM_001773.2 | 1488 | UTR 3 | NP_001764.1 |