Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATTTATAAGCACAATCTTTGGAA[C/T]GCTCTATAGTTTCAAAAAGTGTTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 607055 | ||||||||||||||||||||
Literature Links: |
TFB2M PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TFB2M - transcription factor B2, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022366.2 | Intron | NP_071761.1 | ||||
XM_011544248.2 | Intron | XP_011542550.1 | ||||
XM_017002055.1 | Intron | XP_016857544.1 |