Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGAAGAAGTTGATGCTATAGGT[A/G]ATGGTGCGCATGGTCACGGGGTGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 607348 MIM: 606162 | ||||||||||||||||||||
Literature Links: |
HES5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HES5 - hes family bHLH transcription factor 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PANK4 - pantothenate kinase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018216.2 | 1015 | Silent Mutation | ATC,ATT | I,I 344 | NP_060686.2 | |
XM_011541708.2 | 1015 | Silent Mutation | ATC,ATT | I,I 336 | XP_011540010.2 | |
XM_017001633.1 | 1015 | Silent Mutation | ATC,ATT | I,I 189 | XP_016857122.1 |