Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGGAGGTGTTTGGTGATGACTCT[C/G]AGATCTCTAAAGAATCATCAGGAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 607301 | ||||||||||||||||||||
Literature Links: |
PRPF3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PRPF3 - pre-mRNA processing factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004698.2 | 400 | Missense Mutation | NP_004689.1 | |||
XM_011510129.1 | 400 | Intron | XP_011508431.1 | |||
XM_011510130.2 | 400 | Intron | XP_011508432.1 | |||
XM_011510131.2 | 400 | Missense Mutation | CAG,GAG | Q,E 100 | XP_011508433.1 | |
XM_011510132.2 | 400 | Missense Mutation | CAG,GAG | Q,E 100 | XP_011508434.1 | |
XM_017002790.1 | 400 | Intron | XP_016858279.1 | |||
XM_017002791.1 | 400 | Missense Mutation | CAG,GAG | Q,E 100 | XP_016858280.1 |