Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTATAGGCTCATTCATTTAGCCAG[A/G]CACTCATGCCTTTGTTCTTCAAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611350 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
INTS7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INTS7 - integrator complex subunit 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199809.1 | Intron | NP_001186738.1 | ||||
NM_001199811.1 | Intron | NP_001186740.1 | ||||
NM_001199812.1 | Intron | NP_001186741.1 | ||||
NM_015434.3 | Intron | NP_056249.1 | ||||
XM_011509396.2 | Intron | XP_011507698.1 | ||||
XM_017000962.1 | Intron | XP_016856451.1 |