Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGTGGAAGGCTGGGGTCAGCATC[C/G]GTCGATGCTGGAACCATGTCTGCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 601310 | ||||||||||||||||||||
Literature Links: |
CYP4A11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CYP4A11 - cytochrome P450 family 4 subfamily A member 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000778.3 | Intron | NP_000769.2 | ||||
NM_001319155.1 | Intron | NP_001306084.1 | ||||
XM_005270539.1 | Intron | XP_005270596.1 | ||||
XM_017000464.1 | Intron | XP_016855953.1 | ||||
XM_017000465.1 | Intron | XP_016855954.1 |