Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATCTGTTTATAAATCCCTTTCAC[C/G]GAAACAGCATTAGCGACCTGTAAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602234 MIM: 609444 | ||||||||||||||||||||
Literature Links: |
CASP9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CASP9 - caspase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001229.4 | 971 | Silent Mutation | TCC,TCG | S,S 392 | NP_001220.2 | |
NM_001278054.1 | 971 | Silent Mutation | TCC,TCG | S,S 242 | NP_001264983.1 | |
NM_032996.3 | 971 | Silent Mutation | TCC,TCG | S,S 309 | NP_127463.2 | |
XM_005246014.2 | 971 | Silent Mutation | TCC,TCG | S,S 309 | XP_005246071.1 | |
XM_011542273.2 | 971 | Intron | XP_011540575.1 |
CELA2B - chymotrypsin like elastase family member 2B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |