Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCCCAGGCAGCCATGGACGAGTG[C/T]ACGGACGAGGGCGGGCGGCCGCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 150290 | ||||||||||||||||||||
Literature Links: |
LAMC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LAMC1 - laminin subunit gamma 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002293.3 | 377 | Silent Mutation | TGC,TGT | C,C 40 | NP_002284.3 |