Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGATTCGAGCCAGCTCCTCGCTCC[A/G]TTGCTGGTCCTATGAGGGAAATGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 608854 | ||||||||||||||||||||
Literature Links: |
OSCP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OSCP1 - organic solute carrier partner 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145047.4 | 1149 | Missense Mutation | CGG,TGG | R,W 345 | NP_659484.4 | |
NM_206837.2 | 1149 | Intron | NP_996668.1 | |||
XM_005270461.1 | 1149 | Missense Mutation | CGG,TGG | R,W 355 | XP_005270518.1 | |
XM_005270462.1 | 1149 | UTR 3 | XP_005270519.1 | |||
XM_005270463.3 | 1149 | Intron | XP_005270520.1 | |||
XM_011540680.2 | 1149 | Missense Mutation | CGG,TGG | R,W 327 | XP_011538982.1 | |
XM_011540681.2 | 1149 | Missense Mutation | CGG,TGG | R,W 317 | XP_011538983.1 | |
XM_017000303.1 | 1149 | UTR 3 | XP_016855792.1 | |||
XM_017000304.1 | 1149 | Intron | XP_016855793.1 |