Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGTACTTACAGCACTGAAGTTG[A/G]CAAAATTGCTTGGATCAGCCTCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600051 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPS15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EPS15 - epidermal growth factor receptor pathway substrate 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159969.1 | 2564 | Missense Mutation | GCC,GTC | A,V 530 | NP_001153441.1 | |
NM_001981.2 | 2564 | Missense Mutation | GCC,GTC | A,V 844 | NP_001972.1 | |
XM_005270618.2 | 2564 | Missense Mutation | GCC,GTC | A,V 814 | XP_005270675.1 | |
XM_017000615.1 | 2564 | Missense Mutation | GCC,GTC | A,V 881 | XP_016856104.1 | |
XM_017000616.1 | 2564 | Missense Mutation | GCC,GTC | A,V 813 | XP_016856105.1 | |
XM_017000617.1 | 2564 | Missense Mutation | GCC,GTC | A,V 776 | XP_016856106.1 | |
XM_017000618.1 | 2564 | Missense Mutation | GCC,GTC | A,V 746 | XP_016856107.1 | |
XM_017000619.1 | 2564 | Missense Mutation | GCC,GTC | A,V 656 | XP_016856108.1 | |
XM_017000620.1 | 2564 | Missense Mutation | GCC,GTC | A,V 606 | XP_016856109.1 |