Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATATACTGTCGAGACAGATGAGT[A/G]AATATCCATCTGGTGAGAGAGTACG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 134371 | ||||||||||||||||||||
Literature Links: |
CFHR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CFHR1 - complement factor H related 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002113.2 | 607 | Missense Mutation | AAA,GAA | K,E 165 | NP_002104.2 | |
XM_011509457.2 | 607 | Intron | XP_011507759.1 |