Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGGCTGGGGGGCCGTGGCCAGCC[C/T]AAGGATGAGGTGGACTGGTAGGTAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 616793 | ||||||||||||||||||||
Literature Links: |
PLA2G2F PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLA2G2F - phospholipase A2 group IIF | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022819.3 | 388 | Silent Mutation | CCC,CCT | P,P 99 | NP_073730.3 | |
XM_011541955.1 | 388 | Silent Mutation | CCC,CCT | P,P 99 | XP_011540257.1 | |
XM_011541956.1 | 388 | Silent Mutation | CCC,CCT | P,P 99 | XP_011540258.1 |