Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACGCAAAGGACTTACACAAAGGGA[C/T]CTGAGTCACTAAATGTGAAATACTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607378 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SERBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SERBP1 - SERPINE1 mRNA binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018067.1 | 5795 | UTR 3 | NP_001018077.1 | |||
NM_001018068.1 | 5795 | UTR 3 | NP_001018078.1 | |||
NM_001018069.1 | 5795 | UTR 3 | NP_001018079.1 | |||
NM_015640.3 | 5795 | UTR 3 | NP_056455.3 |