Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAATATACAATTCTTTTCAACTTC[C/T]CCATGTGACTGGTATAAGGAACCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609132 MIM: 601422 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KDM1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KDM1A - lysine demethylase 1A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LUZP1 - leucine zipper protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142546.1 | 4825 | UTR 3 | NP_001136018.1 | |||
NM_033631.3 | 4825 | UTR 3 | NP_361013.3 | |||
XM_011542090.2 | 4825 | UTR 3 | XP_011540392.1 | |||
XM_011542091.2 | 4825 | UTR 3 | XP_011540393.1 | |||
XM_017002252.1 | 4825 | UTR 3 | XP_016857741.1 |